Stampa…

Dimethyl citraconate ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Citraconic Acid Dimethyl Ester, (2Z)-Dimethyl 2-methylmaleate

C0364-25G C0364-5G
TCIAC0364-25GEA 151 EUR
TCIAC0364-25G TCIAC0364-5G
Dimethyl citraconate ≥99.0% (by GC)
Dimethyl citraconate
Formula: C₇H₁₀O₄
Peso molecolare: 158,15 g/mol
Punto di ebollizione: 211°C
MDL Number: MFCD00059306
CAS Number: 617-54-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b85df80d8deb927de14fc5769fbcf73e40a711b&vid=3b85df80d8deb927de14fc5769fbcf73e40a711b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR