Stampa…

Cholesterol-n-octanoate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: n-Octanoic Acid Cholesterol Ester, (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl octanoate, Cholesterol n-Octanoate

C0334-25G C0334-5G
TCIAC0334-25GEA 124 EUR
TCIAC0334-25G TCIAC0334-5G
Cholesterol-n-octanoate ≥95.0% (by GC)
Cholesterol-n-octanoate
Formula: C₃₅H₆₀O₂
Peso molecolare: 512,86 g/mol
MDL Number: MFCD00003642
CAS Number: 1182-42-9
EINECS: 214-656-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Mesomorphic range 107.0 to 112.0 deg-C
Specific rotation [a]20/D -24.0 to -27.0 deg (C=5, THF)
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR