Stampa…

Choline hydroxide 48 - 50% in water

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Hydroxyethyltrimethylammonium Hydroxide, Choline

C0326-25G C0326-500G C0326-100G
TCIAC0326-25GEA 16 EUR
TCIAC0326-25G TCIAC0326-500G TCIAC0326-100G
Choline hydroxide 48 - 50% in water
Choline hydroxide
Formula: C₅H₁₅NO₂
Peso molecolare: 121,18 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002831
CAS Number: 123-41-1
EINECS: 204-625-1
UN: 3267
ADR: 8,II
Merck Index: 14,02204

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light red to Green clear liquid
Concentration (Neutralization titration) 46.5 to 50.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR