4-Benzoylphenyl methacrylate ≥98.0% (by GC)
Fornitore: TCI
B6151-25G
B6151-5G
TCIAB6151-25GEA
121
EUR
TCIAB6151-25G
TCIAB6151-5G
4-Benzoylphenyl methacrylate ≥98.0% (by GC)
4-Benzoylphenyl methacrylate
Formula:
C₁₇H₁₄O₃ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD30470334 CAS Number: 56467-43-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...