Stampa…

4-Bromospiro[fluorene-9,9'-xanthene] ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Bromospiro[9H-fluorene-9,9'-[9H]xanthene], 4-Bromo-spiro[9H-fluorene-9,9'-[9H]xanthene]

B6128-1G B6128-5G
TCIAB6128-1GEA 59 EUR
TCIAB6128-1G TCIAB6128-5G
4-Bromospiro[fluorene-9,9'-xanthene] ≥98.0% (by GC)
4-Bromospiro[fluorene-9,9'-xanthene]
Formula: C₂₅H₁₅BrO
Peso molecolare: 411,30 g/mol
MDL Number: MFCD32065723
CAS Number: 1609484-45-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR