Stampa…

Barium trifluoromethanesulphonate ≥96.0% (W)

Fornitore: TCI
Pericolo

B6120-25G B6120-5G
TCIAB6120-25GEA 175 EUR
TCIAB6120-25G TCIAB6120-5G
Barium trifluoromethanesulphonate ≥96.0% (W)
Barium trifluoromethanesulphonate
Formula: C₂BaF₆O₆S₂
Punto di fusione: 370°C
MDL Number: MFCD00143644
CAS Number: 2794-60-7
EINECS: 220-526-6
UN: 1564
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR