Stampa…

1-(Boc-amino)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B6086-1G B6086-5G
TCIAB6086-1GEA 29 EUR
TCIAB6086-1G TCIAB6086-5G
1-(Boc-amino)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Boc-amino)cyclobutanecarboxylic acid
Formula: C₁₀H₁₇NO₄
MDL Number: MFCD02682623
CAS Number: 120728-10-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR