Stampa…

4-Bromo-7-azaindole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Bromo-7-azaindole, 4-Bromo-1H-pyrrolo[2,3-b]pyridine

B6078-1G B6078-5G
TCIAB6078-1GEA 62 EUR
TCIAB6078-1G TCIAB6078-5G
4-Bromo-7-azaindole ≥98.0% (by GC, titration analysis)
4-Bromo-7-azaindole
Formula: C₇H₅BrN₂
Peso molecolare: 197,04 g/mol
MDL Number: MFCD08272233
CAS Number: 348640-06-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 177.0 to 181.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR