Stampa…

[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

B6032-200MG B6032-1G
TCIAB6032-200MGEA 30 EUR
TCIAB6032-200MG TCIAB6032-1G
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid (contains varying amounts of Anhydride)
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid
Formula: C₁₂H₁₅BO₆
MDL Number: MFCD11867795
CAS Number: 932378-94-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 and le 107.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR