Stampa…

2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl, 3-Biphenylboronic acid Pinacol ester

B6023-1G B6023-5G
TCIAB6023-1GEA 26 EUR
TCIAB6023-1G TCIAB6023-5G
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₈H₂₁BO₂
Peso molecolare: 280,17 g/mol
MDL Number: MFCD18452169
CAS Number: 912844-88-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR