tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
tert-Butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate
B6012-200MG
B6012-1G
TCIAB6012-200MGEA
26
EUR
TCIAB6012-200MG
TCIAB6012-1G
tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate ≥98.0% (by GC)
tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate
Formula:
C₁₂H₁₉NO₄ Peso molecolare: 241,29 g/mol |
MDL Number:
MFCD16140210 CAS Number: 1002355-96-5 |
Risultati del test di specificazione
Appearance | White to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...