2-(tert-Butylamino)ethyl methacrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methacrylic acid 2-(tert-Butylamino)ethyl ester, 2-(tert-Butylamino)ethyl Methacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
B6009-25ML
B6009-500ML
TCIAB6009-25MLEA
16
EUR
TCIAB6009-25ML
TCIAB6009-500ML
2-(tert-Butylamino)ethyl methacrylate ≥98.0% (by GC) stabilizzato
2-(tert-Butylamino)ethyl methacrylate
Formula:
C₁₀H₁₉NO₂ Peso molecolare: 185,27 g/mol Punto di ebollizione: 105°C Flash Pt: 71°C |
MDL Number:
MFCD00014997 CAS Number: 3775-90-4 EINECS: 223-228-4 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...