3-tert-Butylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₅BO₂ Punto di fusione: 221°C |
MDL Number:
MFCD11110647 CAS Number: 560132-24-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 98.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...