Stampa…

2-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

B6000-200MG B6000-1G
TCIAB6000-200MGEA 58 EUR
TCIAB6000-200MG TCIAB6000-1G
2-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
2-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formula: C₂₄H₁₇Br
Peso molecolare: 385,30 g/mol
MDL Number: MFCD32062871
CAS Number: 1890136-54-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR