Stampa…
2-Bromo-7-chloro-9,9-dimethyl-9H-fluorene ≥95.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
B5988-25G B5988-5G
TCIAB5988-25GEA 475 EUR
TCIAB5988-25G TCIAB5988-5G
2-Bromo-7-chloro-9,9-dimethyl-9H-fluorene ≥95.0% (by GC)
2-Bromo-7-chloro-9,9-dimethyl-9H-fluorene
Formula: C₁₅H₁₂BrCl
Peso molecolare: 307,62 g/mol
CAS Number: 605630-37-3
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 155.0 to 159.0 deg-C