Stampa…
3'-Bromo[1,1'-biphenyl]-4-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 4-(3-Bromophenyl)benzonitrile
B5965-1G B5965-5G
TCIAB5965-1GEA 65 EUR
TCIAB5965-1G TCIAB5965-5G
3'-Bromo[1,1'-biphenyl]-4-carbonitrile ≥98.0% (by GC)
3'-Bromo[1,1'-biphenyl]-4-carbonitrile
Formula: C₁₃H₈BrN
Peso molecolare: 258,12 g/mol
CAS Number: 160521-46-0
Visualizza i codici
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C