3'-Bromo[1,1'-biphenyl]-3-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(3-Bromophenyl)benzonitrile
B5964-1G
B5964-5G
TCIAB5964-1GEA
86
EUR
TCIAB5964-1G
TCIAB5964-5G
3'-Bromo[1,1'-biphenyl]-3-carbonitrile ≥98.0% (by GC)
3'-Bromo[1,1'-biphenyl]-3-carbonitrile
Formula:
C₁₃H₈BrN Peso molecolare: 258,12 g/mol |
CAS Number:
1040141-34-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...