Stampa…

5-Bromobenzo[b]naphtho[1,2-d]thiophene ≥97.0% (by GC)

Fornitore: TCI EUROPE

B5940-1G B5940-5G
TCIAB5940-1GEA 119 EUR
TCIAB5940-1G TCIAB5940-5G
5-Bromobenzo[b]naphtho[1,2-d]thiophene ≥97.0% (by GC)
5-Bromobenzo[b]naphtho[1,2-d]thiophene
Formula: C₁₆H₉BrS
Peso molecolare: 313,21 g/mol
CAS Number: 189097-35-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR