Stampa…

5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-triphenylmethyl-2H-tetrazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-trityl-2H-tetrazole

B5933-1G B5933-5G
TCIAB5933-1GEA 67 EUR
TCIAB5933-1G TCIAB5933-5G
5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-triphenylmethyl-2H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-triphenylmethyl-2H-tetrazole
Formula: C₃₃H₂₅BrN₄
Peso molecolare: 557,50 g/mol
Punto di fusione: 137°C
MDL Number: MFCD08274767
CAS Number: 133051-88-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR