Stampa…

1-Boc-3,3-difluoro-4,4-(dihydroxy)piperidine ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(tert-Butoxycarbonyl)-3,3-difluoro-4,4-dihydroxypiperidine, 1-Boc-3,3-difluoro-4,4-dihydroxypiperidine, 3,3-Difluoro-4,4-dihydroxypiperidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 3,3-Difluoro-4,4-dihydroxypiperidine-1-carboxylate

B5931-100MG B5931-500MG
TCIAB5931-100MGEA 52 EUR
TCIAB5931-100MG TCIAB5931-500MG
1-Boc-3,3-difluoro-4,4-(dihydroxy)piperidine ≥97.0% (by GC)
1-Boc-3,3-difluoro-4,4-(dihydroxy)piperidine
Formula: C₁₀H₁₇F₂NO₄
Peso molecolare: 253,25 g/mol
MDL Number: MFCD18072968
CAS Number: 1067914-83-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR