2-Benzyloxy-5-bromopyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Synonyms:
2-(Benzyloxy)-5-bromopyridine
Formula:
C₁₂H₁₀BrNO Punto di ebollizione: 185°C |
MDL Number:
MFCD09037462 CAS Number: 83664-33-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...