Stampa…

2-(Benzyloxy)-1-ethanamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: 2-(Benzyloxy)ethanamine, 2-(Benzyloxy)ethylamine

B5880-1G B5880-5G
TCIAB5880-1GEA 32 EUR
TCIAB5880-1G TCIAB5880-5G
2-(Benzyloxy)-1-ethanamine ≥98.0% (by GC, titration analysis)
2-(Benzyloxy)-1-ethanamine
Formula: C₉H₁₃NO
Punto di ebollizione: 100°C
MDL Number: MFCD00235180
CAS Number: 38336-04-8
UN: 2735
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR