Benzyloxytrimethylsilane ≥95.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₆OSi Punto di ebollizione: 212°C |
MDL Number:
MFCD00134529 CAS Number: 14642-79-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light red clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...