4-(3-Buten-1-yl)-4'-methylbiphenyl ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₇H₁₈ |
CAS Number:
117713-14-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...