Stampa…

(3-(Bromomethyl)-1-(p-toluenesulphonyl)azetidin-3-yl)methanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: [3-(Bromomethyl)-1-(p-toluenesulphonyl)azetidin-3-yl]methanol, [3-(Bromomethyl)-1-tosylazetidin-3-yl]methanol

B5852-200MG B5852-1G
TCIAB5852-200MGEA 65 EUR
TCIAB5852-200MG TCIAB5852-1G
(3-(Bromomethyl)-1-(p-toluenesulphonyl)azetidin-3-yl)methanol ≥98.0% (by GC)
(3-(Bromomethyl)-1-(p-toluenesulphonyl)azetidin-3-yl)methanol
Formula: C₁₂H₁₆BrNO₃S
Peso molecolare: 334,24 g/mol
MDL Number: MFCD18782897
CAS Number: 1041026-55-4

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR