6-Bromochroman-4-one ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
6-Bromo-4-chromanone
B5843-1G
B5843-5G
TCIAB5843-1GEA
24
EUR
TCIAB5843-1G
TCIAB5843-5G
6-Bromochroman-4-one ≥98.0% (by GC)
6-Bromochroman-4-one
Formula:
C₉H₇BrO₂ Peso molecolare: 227,0547 g/mol |
MDL Number:
MFCD00546803 CAS Number: 49660-57-3 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...