2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Bromospiro[fluorene-9,9'-xanthene]
B5842-1G
B5842-5G
TCIAB5842-1GEA
37
EUR
TCIAB5842-1G
TCIAB5842-5G
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene]
Formula:
C₂₅H₁₅BrO Peso molecolare: 411,30 g/mol |
MDL Number:
MFCD29050113 CAS Number: 899422-06-1 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...