Stampa…

(±)-1-tert-Butoxy-2-propanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-2-(tert-butoxy)ethanol, (±)-Monopropylene glycol tert-butyl ether, (±)-Propylene glycol mono-tert-butyl ether, Propylene glycol 1-tert-Butyl ether, ARCOSOLV® PTB, 1-(tert-Butoxy)-2-propanol, 1-tert-Butoxy-2-propanol

B5840-1ML B5840-5ML
TCIAB5840-1MLEA 102 EUR
TCIAB5840-1ML TCIAB5840-5ML
(±)-1-tert-Butoxy-2-propanol ≥98.0% (by GC)
(±)-1-tert-Butoxy-2-propanol
Formula: C₇H₁₆O₂
Peso molecolare: 132,2 g/mol
Punto di ebollizione: 152°C
Punto di fusione: -56°C
Flash Pt: 44°C
MDL Number: MFCD00077674
CAS Number: 57018-52-7
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR