Stampa…

Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylic acid Benzyl ester, N-Cbz-7-oxa-3-azabicyclo[4.1.0]heptane

B5831-200MG B5831-1G
TCIAB5831-200MGEA 40 EUR
TCIAB5831-200MG TCIAB5831-1G
Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥97.0% (by GC)
Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
Formula: C₁₃H₁₅NO₃
Peso molecolare: 233,27 g/mol
Punto di ebollizione: 146°C
MDL Number: MFCD12407051
CAS Number: 66207-08-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR