2-(4-Bromophenyl)-4-phenylquinazoline ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
Formula:
C₂₀H₁₃BrN₂ |
MDL Number:
MFCD27979424 CAS Number: 540466-42-0 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...