4-Bromo-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
B5828-25G
B5828-5G
TCIAB5828-25GEA
270
EUR
TCIAB5828-25G
TCIAB5828-5G
4-Bromo-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Bromo-2-(trifluoromethyl)benzonitrile
Formula:
C₈H₃BrF₃N Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD06659466 CAS Number: 191165-13-6 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...