Stampa…

1-Benzylpiperidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B5826-1G B5826-5G
TCIAB5826-1GEA 80 EUR
TCIAB5826-1G TCIAB5826-5G
1-Benzylpiperidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Benzylpiperidine-4-carboxylic acid
Formula: C₁₃H₁₇NO₂
Peso molecolare: 219,28 g/mol
MDL Number: MFCD03371463
CAS Number: 10315-07-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR