tert-Butyl methyl(2-oxoethyl)carbamate ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
tert-Butyl methyll(2-oxoethyl)carbamate, Methyl(2-oxoethyl)carbamic acid tert-butyl ester, N-Boc-(methylamino)acetaldehyde, N-tert-Butoxycarbonyl-(methylamino)acetaldehyde
B5825-1G
B5825-5G
TCIAB5825-1GEA
55
EUR
TCIAB5825-1G
TCIAB5825-5G
tert-Butyl methyl(2-oxoethyl)carbamate ≥96.0% (by GC)
tert-Butyl methyl(2-oxoethyl)carbamate
Formula:
C₈H₁₅NO₃ Peso molecolare: 173,21 g/mol Punto di ebollizione: 89°C |
MDL Number:
MFCD08064267 CAS Number: 123387-72-4 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Acid Value | max. 5.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...