2-Bromobenzo[b]naphtho[2,3-d]furan ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Bromonaphtho[2,3-b]benzofuran
B5805-1G
B5805-5G
TCIAB5805-1GEA
42
EUR
TCIAB5805-1G
TCIAB5805-5G
2-Bromobenzo[b]naphtho[2,3-d]furan ≥98.0% (by GC)
2-Bromobenzo[b]naphtho[2,3-d]furan
Formula:
C₁₆H₉BrO Peso molecolare: 297,15 g/mol |
MDL Number:
MFCD30187285 CAS Number: 1627917-16-1 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...