Stampa…

2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5803-1G B5803-5G
TCIAB5803-1GEA 57 EUR
TCIAB5803-1G TCIAB5803-5G
2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)
2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-phenyl-1,3,5-triazine
Formula: C₂₁H₁₄ClN₃
Peso molecolare: 343,81 g/mol
MDL Number: MFCD30536776
CAS Number: 1689576-03-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR