Stampa…

2-(Bromomethyl)-5-(trifluoromethyl)furan ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

B5800-200MG B5800-1G
TCIAB5800-200MGEA 33 EUR
TCIAB5800-200MG TCIAB5800-1G
2-(Bromomethyl)-5-(trifluoromethyl)furan ≥97.0% (by GC)
2-(Bromomethyl)-5-(trifluoromethyl)furan
Formula: C₆H₄BrF₃O
Peso molecolare: 228,9946 g/mol
MDL Number: MFCD03086219
CAS Number: 17515-77-4
UN: 2920
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR