2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
B5792-1G
B5792-5G
TCIAB5792-1GEA
44
EUR
TCIAB5792-1G
TCIAB5792-5G
2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine
Formula:
C₂₁H₁₄ClN₃ Peso molecolare: 343,81 g/mol |
MDL Number:
MFCD27979856 CAS Number: 1472062-94-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 163.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...