Bismuth(III) iodide, anidro ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Bismuth triiodide
B5787-5G
B5787-25G
TCIAB5787-5GEA
33
EUR
TCIAB5787-5G
TCIAB5787-25G
Bismuth(III) iodide, anidro ≥98.0% (by titrimetric analysis)
Bismuth(III) iodide
Formula:
BiI₃ Peso molecolare: 589,69 g/mol Punto di ebollizione: 500°C Punto di fusione: 408°C |
MDL Number:
MFCD00010894 CAS Number: 7787-64-6 EINECS: 232-127-4 UN: 3260 ADR: 8,III Merck Index: 14,01269 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...