Bis(3-sulpho-N-succinimidyl) Glutarate disodium salt ≥95.0% (by total nitrogen basis)
Fornitore: TCI
Synonyms:
Pentanedioic acid 1,5-Bis(2,5-dioxo-3-sulpho-1-pyrrolidinyl) ester sodium salt, 1,5-Bis(2,5-dioxo-3-sulpho-1-pyrrolidinyl) Pentanedioate sodium salt, Sodium 1,1'-[Glutaroylbis(oxy)]bis(2,5-dioxopyrrolidine-3-sulphonate), Glutaric acid bis(3-sulpho-N-succinimidyl) disodium salt, sulpho-DSG 2Na
Formula:
C₁₃H₁₂N₂Na₂O₁₄S₂ Peso molecolare: 530.34 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD28358733 CAS Number: 881415-72-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...