SB-366791 ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-Chloro-N-(3-methoxyphenyl)cinnamide, 3-(4-Chlorophenyl)-N-(3-methoxyphenyl)acrylamide, N-(3-Methoxyphenyl)-4-chlorocinnamide
B5762-100MG
B5762-25MG
TCIAB5762-100MGEA
180
EUR
TCIAB5762-100MG
TCIAB5762-25MG
SB-366791 ≥98.0% (by GC)
SB-366791
Formula:
C₁₆H₁₄ClNO₂ Peso molecolare: 287,743 g/mol |
MDL Number:
MFCD01033818 CAS Number: 472981-92-3 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...