Stampa…

1-Bromo-2-chloro-4-iodobenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5742-1G B5742-5G
TCIAB5742-1GEA 16 EUR
TCIAB5742-1G TCIAB5742-5G
1-Bromo-2-chloro-4-iodobenzene ≥98.0% (by GC)
1-Bromo-2-chloro-4-iodobenzene
Formula: C₆H₃BrCl
Peso molecolare: 317,3500 g/mol
MDL Number: MFCD04973904
CAS Number: 535934-25-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR