Stampa…

2-Bromoresorcinol ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromobenzene-1,3-diol

B5737-1G B5737-5G
TCIAB5737-1GEA 24 EUR
TCIAB5737-1G TCIAB5737-5G
2-Bromoresorcinol ≥98.0% (by titrimetric analysis)
2-Bromoresorcinol
Formula: C₆H₅BrO₂
Peso molecolare: 189,008 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD07779241
CAS Number: 6751-75-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystaline
Purity (GC) min. 97.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR