Stampa…
tert-Butylphosphonic dichloride ≥98.0% (by GC)
Fornitore: TCI
Pericolo
B5729-1G B5729-5G
TCIAB5729-1GEA 39 EUR
TCIAB5729-1G TCIAB5729-5G
tert-Butylphosphonic dichloride ≥98.0% (by GC)
tert-Butylphosphonic dichloride
Formula: C₄H₉Cl₂OP
Punto di ebollizione: 110°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00012289
CAS Number: 4707-95-3
UN: 3261
ADR: 8,II
Visualizza i codici
Appearance White to Light yellow crystal to lump, wax
Purity (GC) min. 98.0 %
Melting point 120.0 to 125.0 deg-C