Stampa…

9-([1,1'-Biphenyl]-4-yl)anthracene ≥98.0% (by GC)

Fornitore: TCI EUROPE

B5717-1G B5717-5G
TCIAB5717-1GEA 38 EUR
TCIAB5717-1G TCIAB5717-5G
9-([1,1'-Biphenyl]-4-yl)anthracene ≥98.0% (by GC)
9-([1,1'-Biphenyl]-4-yl)anthracene
Formula: C₂₆H₁₈
Peso molecolare: 330,43 g/mol
MDL Number: MFCD27979543
CAS Number: 323195-31-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 221.0 to 225.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR