Brequinar ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
6-Fluoro-2-(2'-fluoro-[1,1'-biphenyl]-4-yl)-3-methylquinoline-4-carboxylic acid
B5707-20MG
B5707-100MG
TCIAB5707-20MGEA
78
EUR
TCIAB5707-20MG
TCIAB5707-100MG
Brequinar ≥98.0% (by HPLC)
Brequinar
Formula:
C₂₃H₁₅F₂NO₂ Peso molecolare: 375,37 g/mol Punto di fusione: 317°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00866437 CAS Number: 96187-53-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 3.50 to 4.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...