Stampa…

2-(Boc-amino)-4-methylpyridine ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-[(tert-Butoxycarbonyl)amino]-4-methylpyridine, 2-(Boc-amino)-4-methylpyridine, (4-Methylpyridin-2-yl)carbamic acid tert-Butyl ester, tert-Butyl (4-Methylpyridin-2-yl)carbamate

B5706-1G B5706-5G
TCIAB5706-1GEA 119 EUR
TCIAB5706-1G TCIAB5706-5G
2-(Boc-amino)-4-methylpyridine ≥97.0% (by GC)
2-(Boc-amino)-4-methylpyridine
Formula: C₁₁H₁₆N₂O₂
Peso molecolare: 208,261 g/mol
MDL Number: MFCD07776933
CAS Number: 90101-20-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 115.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR