Stampa…

N-Benzyl-2,2,2-trifluoroacetamide ≥95.0% (by GC)

Fornitore: TCI

B5705-1G B5705-5G
TCIAB5705-1GEA 113 EUR
TCIAB5705-1G TCIAB5705-5G
N-Benzyl-2,2,2-trifluoroacetamide ≥95.0% (by GC)
N-Benzyl-2,2,2-trifluoroacetamide
Formula: C₉H₈F₃NO
MDL Number: MFCD00188542
CAS Number: 7387-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54bdf82a8495901dca150558f74a230d386b208&vid=f54bdf82a8495901dca150558f74a230d386b208&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR