2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2,2'-Biphenyldiboronic acid bis(pinacol) ester
B5703-200MG
B5703-1G
TCIAB5703-200MGEA
60
EUR
TCIAB5703-200MG
TCIAB5703-1G
2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl ≥98.0% (by GC, titration analysis)
2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl
Formula:
C₂₄H₃₂B₂O₄ Peso molecolare: 406,14 g/mol |
MDL Number:
MFCD30729880 CAS Number: 398128-09-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...