DTME (dithio-bis-maleimidoethane) ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
DTME
B5699-100MG
B5699-25MG
TCIAB5699-100MGEA
375
EUR
TCIAB5699-100MG
TCIAB5699-25MG
DTME (dithio-bis-maleimidoethane) ≥98.0% (by HPLC, total nitrogen)
DTME (dithio-bis-maleimidoethane)
Formula:
C₁₂H₁₂N₂O₄S₂ Peso molecolare: 312,36 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD01863385 CAS Number: 71865-37-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...