3-Bromo-4-iodoanisole ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Bromo-1-iodo-4-methoxybenzene
B5697-1G
B5697-5G
TCIAB5697-1GEA
55
EUR
TCIAB5697-1G
TCIAB5697-5G
3-Bromo-4-iodoanisole ≥97.0% (by GC)
3-Bromo-4-iodoanisole
Formula:
C₇H₆BrIO Peso molecolare: 312,9304 g/mol |
MDL Number:
MFCD12547848 CAS Number: 466639-53-2 |
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...